missing translation for 'onlineSavingsMsg'
Learn More

DL-Dithiothreitol, 99.5%, MP Biomedicals™

£51.65 - £564.00

Chemical Identifiers

CAS 27565-41-9
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00004877,MFCD00064305
InChI Key VHJLVAABSRFDPM-UHFFFAOYNA-N
Synonym Cleland's reagent, DTT
IUPAC Name 1,4-disulfanylbutane-2,3-diol
SMILES OC(CS)C(O)CS
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446531
View Documents
MP Biomedicals
0219482101
1 g
£51.65
1g
Please to purchase this item. Need a web account? Register with us today!
11456531
View Documents
MP Biomedicals
0219482105
5 g
£137.00
5g
Please to purchase this item. Need a web account? Register with us today!
11466531
View Documents
MP Biomedicals
0219482110
10 g
£256.00
10g
Please to purchase this item. Need a web account? Register with us today!
11476531
View Documents
MP Biomedicals
0219482125
25 g
£564.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

27565-41-9
154.24
VHJLVAABSRFDPM-UHFFFAOYNA-N
1,4-disulfanylbutane-2,3-diol
C4H10O2S2
MFCD00004877,MFCD00064305
Cleland's reagent, DTT
OC(CS)C(O)CS

Specifications

27565-41-9
>99%
MFCD00004877,MFCD00064305
VHJLVAABSRFDPM-UHFFFAOYNA-N
1,4-disulfanylbutane-2,3-diol
154.2
Powder
White
C4H10O2S2
Cleland's reagent, DTT
OC(CS)C(O)CS
154.24
Molecular Biology Reagent
DL-Dithiothreitol
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 248-531-9

Recommended Storage : Store at + 4°C

Product Identifier
  • DL-Dithiothreitol, molecular biology reagent

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : DL-Dithiothreitol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.