missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Phenylalanine, 99%

£17.59 - £207.03

Chemical Identifiers

CAS 150-30-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064225
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
PubChem CID 994
ChEBI CHEBI:28044
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11499382
View Documents
Thermo Scientific Chemicals
A10132.14
25 g
£17.59
25g
Please to purchase this item. Need a web account? Register with us today!
11409392
View Documents
Thermo Scientific Chemicals
A10132.22
100 g
£42.81
100g
Please to purchase this item. Need a web account? Register with us today!
11419392
View Documents
Thermo Scientific Chemicals
A10132.36
500 g
£207.03
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Phenylalanine is used to relieve cases of minor depression and other mood issues as well as and physical discomforts and pain. It is also used in acupuncture anesthesia, alcohol withdrawal.

Solubility
Soluble in water. Insoluble in ethyl ether, benzene.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

150-30-1
165.19
COLNVLDHVKWLRT-UHFFFAOYNA-N
994
2-amino-3-phenylpropanoic acid
C9H11NO2
MFCD00064225
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
CHEBI:28044
NC(CC1=CC=CC=C1)C(O)=O

Specifications

∼266°C (decomposition)
C9H11NO2
1910407
dl-phenylalanine, phenylalanine, h-dl-phe-oh, dl-3-phenylalanine, phenylalanin, dlpa, dl-2-amino-3-phenylpropanoic acid, alanine, phenyl-, dl, 2-amino-3-phenylpropionic acid, phenylalanine, dl
COLNVLDHVKWLRT-UHFFFAOYNA-N
2-amino-3-phenylpropanoic acid
994
165.19
DL-Phenylalanine
150-30-1
MFCD00064225
14,7271
Soluble in water. Insoluble in ethyl ether,benzene.
NC(CC1=CC=CC=C1)C(O)=O
165.19
CHEBI:28044
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-756-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.