missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Tryptophan, 98% Product Code.: 10008581

Thermo Scientific Chemicals DL-Tryptophan, 98%

Product Code. 10008581
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10008581

Brand: Thermo Scientific Chemicals 172110250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-12-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064339
InChI Key QIVBCDIJIAJPQS-UHFFFAOYSA-N
Synonym dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .Show MoreShow Less
PubChem CID 1148
ChEBI CHEBI:27897
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Color Beige to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Loss on Drying 0.8% max. (105°C, 3 hrs) (vacuum)
Other Amino Acids (TLC) Not detected
Packaging Plastic Bottle
Beilstein 22, 550
Quantity 25 g
Formula Weight 204.23
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material DL-Tryptophan, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0426003 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number172110250,172111000
Lot NumberA0406415 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number172110250,172111000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Tryptophan, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.