Learn More
Tocris Bioscience™ DMSO, Cell Cryopreserve Grade
Intracellular cryoprotective agent
Brand: Tocris Bioscience™ 7726/70ML
239.40 GBP valid until 2025-04-30
Use promo code "24247" to get your promotional price.
Description
DMSO, Cell Cryopreserve Grade is a cryoprotective agent which penetrates the cell membrane. DMSO, Cell Cryopreserve Grade reduces the osmotic charge of the cells during freezing and thawing and antagonizes the osmotic shock. Furthermore DMSO, Cell Cryopreserve Grade protects the cells from dehydration and shrinking during freezing and prevents the formation of ice crystals.
Specifications
Dimethyl sulfoxide | |
70 mL | |
C2H6OS |
67-68-5 | |
Buffers, Chelators and Reagents | |
0.999 |
Special Offers
-
Multiple Manufacturers
Life Sciences Monthly Promotion
Discover exclusive monthly deals on life sciences products to advance your research and innovation.Promo Code: 24247Expires: 2025-04-30
New Offer Save Up to 52%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.