missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNA Polymerase I (10 U/μL)

DNA Polymerase I is a template-dependent DNA polymerase which catalyzes 5'→3' synthesis of DNA.

Brand:  Thermo Scientific™ EP0041

37.48 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10364630

  • £59.50 / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.

Highlights

  • Incorporates modified nucleotides(e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
  • Active in multiple buffers, including restriction enzyme, PCR, and RT buffers

Applications

  • DNA labeling by nick-translation in conjunction with DNase
  • Second-strand synthesis of cDNA in conjunction with RNase
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA Polymerase I
DNA Polymerase I
500 U
10 U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.