Learn More
Thermo Scientific™ DNA Polymerase I (10 U/μL)
DNA Polymerase I is a template-dependent DNA polymerase which catalyzes 5'→3' synthesis of DNA.
Brand: Thermo Scientific™ EP0042
222.31 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 88
Additional Details : Weight : 1.51950kg
Description
DNA Polymerase I, a template-dependent DNA polymerase, catalyzes 5'→3' synthesis of DNA. The enzyme also exhibits 3'→5' exonuclease (proofreading) activity, 5'→3' exonuclease activity, and ribonuclease H activity.
Highlights
- Incorporates modified nucleotides(e.g. biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides)
- Active in multiple buffers, including restriction enzyme, PCR, and RT buffers
Applications
- DNA labeling by nick-translation in conjunction with DNase
- Second-strand synthesis of cDNA in conjunction with RNase
Specifications
DNA Polymerase I | |
DNA Polymerase I | |
2500 U | |
10 U/μL |
For Research Use Only. Not for use in diagnostic procedures.