missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Retardation Gels (6%), 1.0 mm

Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp.

Brand:  Invitrogen™ EC63655BOX

Code : 50

Additional Details : Weight : 0.68000kg

 View more versions of this product

Product Code. 12000096

  • £186.00 / Pack of 10
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.

Description

Description

Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp. This preparation is good for electrophoretic separation, yet low enough to promote DNA-protein interactions. Novex DNA Retardation Gels are designed to run on the XCell SureLock™ Mini-Cell and are available in 10-, 12-, or 15-well formats.

• DNA fragments ranging from 60-2,500 bp
• Prepared with 0.5X TBE as gel buffer
• Designed to run on the XCell SureLock Mini-Cell
• Available in 10, 12, and 15 well formats
• Manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS

To ensure best possible results, these gels are single use and individually wrapped. Detection is performed with ethidium bromide staining of DNA or, for greater sensitivity, with radiolabeling the DNA or protein.

Formulation
Novex DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS

Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

• Catalog number refers to a single box of 10 gels

Store at 4°C.
XCell SureLock Mini-Cell
1 mm
DNA Retardation Gel
15-well
TBE Hi-Density Sample Buffer (5X)
Wet Ice
DNA Retardation Gels, 15-well
6%
DNA Retardation
60 to 2500 bp
10 Gels/Box
Double-stranded DNA (dsDNA)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers