Learn More
Invitrogen™ DNA Retardation Gels (6%), 1.0 mm
Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp.
£183.00 - £186.00
Specifications
Content And Storage | • Catalog number refers to a single box of 10 gels Store at 4°C. |
---|---|
For Use With (Equipment) | XCell SureLock Mini-Cell |
Gel Percentage | 6% |
GelThickness | 1 mm |
Gel Type | DNA Retardation |
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Wells | Price | Quantity & Availability | ||||
12080086
|
Invitrogen™
EC6365BOX |
DNA Retardation Gels, 10-well | 10-well |
£184.00
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
12090086
|
Invitrogen™
EC63652BOX |
DNA Retardation Gels, 12-well | 12-well |
£183.00
Pack of 10 |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||||
12000096
|
Invitrogen™
EC63655BOX |
DNA Retardation Gels, 15-well | 15-well |
£186.00
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
Description
Novex™ DNA Retardation Gels are 6% polyacrylamide prepared with 0.5X TBE gel buffer, providing clear resolution of fragments used for DNA retardation assays from 60–2500 bp. This preparation is good for electrophoretic separation, yet low enough to promote DNA-protein interactions. Novex DNA Retardation Gels are designed to run on the XCell SureLock™ Mini-Cell and are available in 10-, 12-, or 15-well formats.
• DNA fragments ranging from 60-2,500 bp
• Prepared with 0.5X TBE as gel buffer
• Designed to run on the XCell SureLock Mini-Cell
• Available in 10, 12, and 15 well formats
• Manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS
To ensure best possible results, these gels are single use and individually wrapped. Detection is performed with ethidium bromide staining of DNA or, for greater sensitivity, with radiolabeling the DNA or protein.
Formulation
Novex DNA Retardation Gels are manufactured with high-purity tris base, boric acid, EDTA, acrylamide, bisacrylamide, TEMED, and APS
Recommended buffers
For optimum performance, Novex™ TBE Running Buffer and Novex™ Hi-Density TBE Sample Buffer are strongly recommended for use with these gels. Novex Hi-Density TBE Sample Buffer contains the tracking dyes Bromophenol Blue and Xylene Cyanol as well as the density agent Ficoll™, which yields sharper and straighter bands than conventional density agents.
Order Info
Shipping Condition: Wet Ice
Specifications
• Catalog number refers to a single box of 10 gels Store at 4°C. |
|
6% | |
DNA Retardation | |
60 to 2500 bp | |
TBE Hi-Density Sample Buffer (5X) | |
Wet Ice |
XCell SureLock Mini-Cell | |
1 mm | |
DNA Retardation Gel | |
10 Gels/Box | |
Double-stranded DNA (dsDNA) |