missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17599-1-AP-20UL
This item is not returnable.
View return policy
Specifications
DNAJC16 | |
Polyclonal | |
Unconjugated | |
DNAJC16 | |
DNAJC16, KIAA0962 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214063, 23341, 362652 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5FVM7, Q80TN4, Q9Y2G8 | |
Dnajc16 | |
DNAJC16 Fusion Protein Ag11637 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction