Learn More
Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals
£62.99 - £1356.00
Chemical Identifiers
CAS | 17786-31-1 |
---|---|
Molecular Formula | C12Co4O12 |
Molecular Weight (g/mol) | 571.853 |
MDL Number | MFCD00064698 |
InChI Key | YMFAWOSEDSLYSZ-UHFFFAOYSA-N |
Synonym | dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min |
PubChem CID | 6096999 |
IUPAC Name | carbon monoxide;cobalt |
SMILES | [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]Show More |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11342138
|
Thermo Scientific Alfa Aesar
013055.03 |
1 g | N/A | ||||||
11322138
|
Thermo Scientific Alfa Aesar
013055.06 |
5 g |
£383.91
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332138
|
Thermo Scientific Alfa Aesar
013055.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17786-31-1 | |
571.853 | |
YMFAWOSEDSLYSZ-UHFFFAOYSA-N | |
6096999 | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]Show More |
C12Co4O12 | |
MFCD00064698 | |
dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min | |
carbon monoxide;cobalt |
Specifications
60°C (dec.) | |
Black | |
C12Co4O12 | |
MFCD00064698 | |
dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co] | |
571.853 | |
571.86 | |
Air Sensitive | |
Dodecacarbonyltetracobalt |
17786-31-1 | |
Crystalline | |
Co4(CO)12 | |
UN3179 | |
YMFAWOSEDSLYSZ-UHFFFAOYSA-N | |
carbon monoxide;cobalt | |
6096999 | |
≤98% | |
2.09g/mL |
Safety and Handling
GHS H Statement
H228-H301-H331-H351-H317
Flammable solid.
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P210-P280h-P262-P304+P340-P309-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
missing translation for 'rtecsNumber' : GG3410000
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.