missing translation for 'onlineSavingsMsg'
Learn More
Dodecenylsuccinic acid, Thermo Scientific Chemicals Product Code.: 17261252
compliance-icons

Dodecenylsuccinic acid, Thermo Scientific Chemicals

Product Code. 17261252
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 17261252

Brand: Thermo Scientific Acros 466600010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29658-97-7
Molecular Formula C16H28O4
InChI Key QDCPNGVVOWVKJG-VAWYXSNFNA-N
SMILES CCCCCCCCCC\C=C\C(CC(O)=O)C(O)=O
IUPAC Name 2-[(1E)-dodec-1-en-1-yl]butanedioic acid
Molecular Weight (g/mol) 284.40
Formula Weight 284.39
Quantity 1 g
Chemical Name or Material Dodecenylsuccinic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464153 Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number466600010
Lot NumberA0457874 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number466600010
Lot NumberA0449632 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number466600010
Lot NumberA0443500 Certificate TypeCertificate of Analysis Date24/06/2022 Catalog Number466600010
Lot NumberA0418591 Certificate TypeCertificate of Analysis Date23/07/2020 Catalog Number466600010
compliance-icons
Product Identifier
  • Dodecenylsuccinic acid
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dodecenylsuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dodecenylsuccinic acid, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.