missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Dog (Beagle) Microsomes
Dog (Beagle) Microsomes
Brand: Gibco™ DGMCPL
Code : 50
Additional Details : Weight : 0.50000kg
Description
Liver microsomes prepared from a pool of male Beagle Dogs.Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Specifications
20 mg/mL | |
Store between -60°C and -85°C | |
Male | |
Microsomes | |
Liver | |
Gibco™ |
Subcellular Fractions | |
Pooled | |
Dry Ice | |
Dog/Canine | |
Frozen | |
0.5 mL |
For Research Use Only. Not for use in diagnostic procedures.