missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSIPET B

Dosipet B is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.

Brand:  KARTELL 8300200

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10335971

  • £384.00 / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket

Description

Description

  • For universal use; ideal for use with reagent of average aggressiveness.
  • It can be used with every water solution type (except those containing hydrofluoric acid), marked by a high degree of convenience and reliability. It is equipped with a 3.3 borosilicate glass cylinder and PTFE piston.
  • Precise and quick volume adjustment using the analogue slider
  • Precise graduated scale to ensure reproducibility of the dispensed volumes
  • No loss of reagent when eliminating air or filling
  • Dispensing without bubbles forming
  • The piston completely transfers the liquid with every movement
  • Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
  • Autoclavable at +121°C for 20 minutes
Specifications

Specifications

± 0.012 mL
Autoclavable
1
2.5 mL
0.05 mL
Dosipet B
White
Borosilicate Glass, PTFE
Analog
0.08 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers