missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL DOSIPET B
Dosipet B is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.
£384.00 - £930.00
Specifications
Autoclavable | Autoclavable |
---|---|
Color | White |
Channels | 1 |
Material | Borosilicate Glass, PTFE |
Display | Analog |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10335971
|
KARTELL
8300200 |
2.5 mL |
£384.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10172711
|
KARTELL
8300500 |
5 mL |
£384.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10011031
|
KARTELL
8301000 |
10 mL |
£384.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10798682
|
KARTELL
8306000 |
100 mL |
£930.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
Description
- For universal use; ideal for use with reagent of average aggressiveness.
- It can be used with every water solution type (except those containing hydrofluoric acid), marked by a high degree of convenience and reliability. It is equipped with a 3.3 borosilicate glass cylinder and PTFE piston.
- Precise and quick volume adjustment using the analogue slider
- Precise graduated scale to ensure reproducibility of the dispensed volumes
- No loss of reagent when eliminating air or filling
- Dispensing without bubbles forming
- The piston completely transfers the liquid with every movement
- Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
- Autoclavable at +121°C for 20 minutes
Specifications
Autoclavable | |
1 | |
Analog |
White | |
Borosilicate Glass, PTFE |