Learn More
Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe
Dose with excellent reproducibility using a pistol grip handle automatic syringe. Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
Brand:  Socorex™ 172.0505
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10385002
  • £120.00 / Each
Estimated Shipment: 05-11-2024
to see stock.
Description
Description
  • Pistol grip handle.
  • Automatic syringe.
  • Without valve system.
  • Luer lock nozzle.
  • Fits either hand.
  • Requires limited effort.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness.
  • Glass barrel protected by PVC sleeve (not autoclavable).
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
Specifications
Specifications
Autoclavable
Syringe, PVC protection sleeve, operating instructions.
0.5 mL
1/ea.
0.5 to 5 mL
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Automatic Syringe, Pistol Grip
Dosys™ Basic 172
<0.5% to <0.2%
Yes