Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
Brand: Metro™ LBTM21S
This item is not returnable.
View return policy
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
- Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Mobile Unit Kit | |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system | |
21 in. | |
53 cm | |
Stainless Steel |
Stainless Steel | |
Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware | |
21 in. | |
53 cm |