missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Double Spatula, Flat, Nickel
Double-ended for versatility. Bochem™ Double Spatula, Flat, Nickel has dual flat ends and is constructed of resistant Nickel (99.5%).
Brand: Bochem™ 3279
Additional Details : Weight : 0.10000kg
Description
- Double-ended.
- Dual flat ends.
- Nickel (99.5%) construction.
Specifications
Spatula | |
20 mm | |
Nickel (99.5%) | |
100 mm |