missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Double Spatula, Nickel

Double-ended for versatility. Bochem™ Double Spatula, Nickel has both a flat end, and a rounded end, and is constructed of resistant Nickel (99.5%).

Brand:  Bochem™ 3110

 View more versions of this product

Product Code. 10098460

  • £36.70 / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Double-ended.
  • Round end and flat end.
  • Nickel (99.5%) construction.
Specifications

Specifications

Spatula
40 mm
Nickel (99.5%)
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers