missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Double Spatula, Nickel
Double-ended for versatility. Bochem™ Double Spatula, Nickel has both a flat end, and a rounded end, and is constructed of resistant Nickel (99.5%).
Brand: Bochem™ 3111
This item is not returnable.
View return policy
Description
- Double-ended.
- Round end and flat end.
- Nickel (99.5%) construction.
Specifications
Spatula | |
45 mm | |
Nickel (99.5%) | |
150 mm |