Learn More
Thermo Scientific Chemicals DuPont Molykote High-Vacuum Grease
Brand:  Thermo Scientific Chemicals 044224.KT
63.97 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 15471977
  • £68.22 / Each
Explore more special offers
Description
Description
Recommended for vacuums reaching between 1 x 10-5 and 1 x 10-6 mm HgDow Corning™ High-Vacuum Grease is used as an anti-seize grease in ground glass joints. It is used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used as an anti-seize grease in ground glass joints. Also used to lubricate synthetic and silicone rubber gaskets. Further, it is used to lubricate plug valves, control valves, flow meter bearings, ceramic plug cocks and fire extinguisher valves. In addition to this, it is used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
1 Ea.
5.3 oz.
Tube
-40°C to +204°C
150 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3167127 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number
Lot Number3167127 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific Chemicals DuPont Molykote High-Vacuum Grease > 1each