Learn More
Gibco™ DPBS (10X), no calcium, no magnesium
GREENER_CHOICE
DPBS (10X), no calcium, no magnesium
Brand:  Gibco™ 14200059
 View more versions of this product
Product Code. 11520486
  • £7.19 / 100mL
Edge
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ DPBS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Sodium Pyruvate
15°to 30°C
DPBS
Mammalian Cell Culture
6 to 7
Liquid
100 mL
10 X
100mL
2500 to 2900 mOsm/kg
For in-vitro diagnostic use