missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ CMS Chips: Hydrogen Sulfide

Suitable for 10 measurements of one gas.

Brand:  Dräger™ 6406050

Code : 50

Additional Details : Weight : 0.02880kg

 View more versions of this product

Product Code. 10138091

  • £73.00 / Each
Estimated Shipment: 21-08-2024
to see stock.

Description

Description

Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications

Specifications

700 to 1100 hpa
1 to 40mg/L
20 to 200 sec.
0°C to 40°C
Hydrogen Sulphide CMS Chip
2 to 50
32°F to 104°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers