Learn More
Dräger™ CMS Chips: Ammonia
Suitable for 10 measurements of one gas
Brand: Dräger™ 6406020
This item is not returnable.
View return policy
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
1 to 40mg/L | |
15 to 50 sec. | |
0°C to 40°C |
Ammonia CMS Chip | |
10 to 150 | |
32°F to 104°F |