missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Base for Schilling Automatic Burette Bottle
For use with Schilling automatic burette. Holds one 500mL plastic bottle with a diameter of 74 to 76mm.
Brand: BRAND™ 23725
This item is not returnable.
View return policy
Description
- For use with Schilling automatic burette
- Holds one 500mL plastic bottle 500ml, dia.74 to 76mm

Specifications
Plastic Bottles Base | |
Schilling automatic buret bottle, 500mL |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Base for Schilling Automatic Burette Bottle > Plastic Bottle Base
Spot an opportunity for improvement?Share a Content Correction