missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EquiPhi29™ DNA Amplification Kit

The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).

Brand:  Thermo Scientific™ A65393

 View more versions of this product

Product Code. 18094070

  • £334.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific EquiPhi29 DNA Amplification Kit includes all required components for fast, sensitive, and efficient isothermal DNA amplification, which includes rolling circle amplification (RCA) and multiple displacement amplification – whole genome amplification (MDA-WGA).

Features of the EquiPhi29 DNA Amplification Kit include:
• Fast DNA amplification in less than two hours
• Sensitivity from 1 fg of input DNA
• Maximal yield of 17 µg amplified DNA
• Compatibility with different types of sample input material (e.g., purified DNA, liquid media culture, agar plate colonies)
• Compatibility with various downstream applications such as DNA sequencing (Sanger sequencing, next-generation sequencing), digestion with restriction enzymes, cell-free DNA enrichment, and cell-free protein expression
• All required components for DNA amplification in RCA or MDA-WGA reactions

The kit contains EquPhi29 DNA Polymerase, a proprietary phi29 DNA polymerase mutant that possesses strong strand displacement activity and enables high thermostability, reaction speed, and product yield and low amplification bias. Also included is an optimized reaction buffer, exo-resistant random primer, pyrophosphatase, and dNTPs. The addition of pyrophosphatase to the reaction mixture enhances DNA amplification and facilitates sample handling.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA Amplification Kit
RCA, MDA-WGA
100 Reactions

• EquiPhi29 DNA Polymerase (0.1 mL at 10 U/μL)
• 10X EquiPhi29 DNA Polymerase Reaction Buffer (1 mL)
• 0.1 M DTT (0.25 mL)
• 500 μM Exo-Resistant Random Primer (0.2 mL)
• 10 mM dNTP Mix (0.2 mL)
• 0.1 U/μL Pyrophosphatase, inorganic (0.1 mL)

Store at -15 to -25°C.

42°C
Up to 2 hours
Liquid
EquiPhi29 DNA Polymerase
Nucleic Acid Analysis, In Vitro Transcription, Cell-free Protein Expression, In Situ Sequencing, SNP Detection
Kit
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2896885 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA65393
Lot Number2982441 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA65393
Lot Number2982423 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberA65393
Lot Number2931668 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA65393
Lot Number2931670 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA65393
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ EquiPhi29™ DNA Amplification Kit > 100 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.