Learn More
Thermo Scientific™ DRAQ5™ Fluorescent Probe Solution (5 mM)
Stain live or fixed cells with this far-red DNA stain.
Brand: Thermo Scientific™ 62252
1558.00 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Stain live or fixed cells with Thermo Scientific™ DRAQ5 Fluorescent Probe, a far-red DNA stain.
Fluorescence cell-based assays, such as flow cytometry, in-cell ELISA, fluorescence microscopy and high-content imaging require a fluorescent label to identify individual cells. When using multiple fluorescent probes to detect different cellular targets or activities, each probe must have a fluorescent spectrum different than the other probes. The blue-fluorescent DNA-binding probes, Hoechst and DAPI are frequently used; however, these probes cannot be used when UV illumination is unavailable or other blue-emitting fluorescent probes are used. Therefore, nuclear probes that emit in a color other than blue is useful for cell identification and counting, and for determining nuclear morphology and DNA content.- Due to its far-red excitation and emission, the DRAQ5 Fluorescent Probe can be multiplexed with many other fluorophores and is ideal for cells expressing green fluorescent protein (GFP) fusion proteins
- Compatible with many existing protocols across a wide range of instrumentation platforms
- Emits in the far-red region, is lipophilic and crosses cell and nuclear membranes in live and fixed cells and tissues for rapid DNA staining
- Stain is water-soluble, supplied ready-to-use and does not require RNase, cell lysis, or a washing step, making it compatible with automation
- Because DNA staining is stoichiometric, the DRAQ5 Fluorescent Probe can be used for DNA content analysis in cell proliferation studies

Specifications
Cell-permeant | |
Store at 4°C. | |
Fluorescence | |
Fluorescence Microscope, High Content Analysis Instrument | |
Cell-Permeant | |
647 nm | |
Pierce™ | |
1 mL | |
Nucleus, Cytosol, Cytoplasm |
Red | |
DRAQ5 Fluorescent Probe Solution (5 mM) | |
Cell Imaging | |
Fluorescent Probe Solution | |
681 nm | |
Liquid | |
> 99% | |
Soluble at working concentrations in water and typical buffers |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
602DR51000 | Certificate of Analysis | 27/01/2024 | 62252 |
601DR51000 | Certificate of Analysis | 25/01/2024 | 62252 |
538DR51000 | Certificate of Analysis | 07/12/2023 | 62252 |
536DR51000 | Certificate of Analysis | 26/07/2023 | 62252 |
535DR51000 | Certificate of Analysis | 20/04/2023 | 62252 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.