Learn More
Thermo Scientific™ DreamTaq™ Buffers (10X)
Thermo Scientific 10X DreamTaq Buffer is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase.
Brand: Thermo Scientific™ B71
15.18 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific DreamTaq Buffer (10X), available in colorless and green formats, is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase. It contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2
In addition, 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes. The density reagent allows direct loading of PCR products on a gel. The blue dye (migrates with 3–5 kb DNA fragments in 1% agarose gel) and the yellow dye (migrates faster than 10 bp DNA fragments in 1% agarose gel) are included for monitoring electrophoresis progress. The dyes have excitation peaks at 424 nm and 615 nm.
Notes
- For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless DreamTaq Buffer.
Specifications
Dream TAQ Green Buffer | |
DreamTaq Hot Start DNA Polymerase, DreamTaq DNA Polymerase | |
DreamTaq Green Buffer | |
Wet or Dry Ice | |
10X |
• 10X DreamTaq Green Buffer (with 20 mM MgCl2), 4 x 1.25 mL Store in a non-frost-free freezer at -20°C. |
|
Nonidet P40 Ethyl phenolpoly(ethyleneglycolether) 9016-45-9 ( <0.9 ), Polyethylene glycol 25322-68-3 ( 7-13 ), Sucrose 57-50-1 ( 10-30 ) | |
4 x 1.25 mL | |
Hot-start PCR | |
Green |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.