Learn More
asecos™ Drum Rack With Sump Pallet Steel
The Drum Rack With Sump Pallet Steel provides safe storage and filling storage of substances hazardous to water of all hazard classes.
Brand:  asecos™ 13246
 View more versions of this product
Product Code. 17806426
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For the storage of oil, paint and lacquer.
  • Safe dispensing due to overhanging collecting sump.
Specifications
Specifications
54.2 gal.
Steel
52.36 in.
52.36 in.
61.81 in.
32.09 in.
306.44 lb.
205 L
Drum Rack
1330.00 mm
1330.00 mm
1570.00 mm
815.00 mm
139 kg