missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Drum Rack With Sump Pallet Steel
The Drum Rack With Sump Pallet Steel provides safe storage and filling storage of substances hazardous to water of all hazard classes.
£606.32 - £929.62
Specifications
Material | Steel |
---|---|
Type | Drum Rack |
Height (English) | 61.81 in. |
Height (Metric) | 1570.00 mm |
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
17856416
|
asecos™
13148 |
66 kg |
£606.32
Each |
Estimated Shipment: 11-11-2024
Log in to see stock. |
|||||
17836406
|
asecos™
13146 |
73 kg |
£720.80
Each |
Estimated Shipment: 11-11-2024
Log in to see stock. |
|||||
17866366
|
asecos™
13089 |
118 kg |
£829.98
Each |
Estimated Shipment: 11-11-2024
Log in to see stock. |
|||||
17806426
|
asecos™
13246 |
139 kg | N/A | ||||||
Description
- For the storage of oil, paint and lacquer.
- Safe dispensing due to overhanging collecting sump.
Specifications
Steel | |
Drum Rack | |
61.81 in. | |
1570.00 mm |