missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Drum Rack With Sump Pallet Steel

The Drum Rack With Sump Pallet Steel provides safe storage and filling storage of substances hazardous to water of all hazard classes.

Brand:  asecos™ 13146

 View more versions of this product

Product Code. 17836406

  • £720.80 / Each
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For the storage of oil, paint and lacquer.
  • Safe dispensing due to overhanging collecting sump.
Specifications

Specifications

17.2 gal.
Steel
29.72 in.
29.72 in.
61.81 in.
25.00 in.
160.94 lb.
65 L
Drum Rack
755.00 mm
755.00 mm
1570.00 mm
635.00 mm
73 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers