Learn More
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone
Excellent for protecting reactions against moisture while equalizing pressure. Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
£21.20 - £29.30
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
10007730
|
Lenz Laborglasinstrumente™
05337001 |
NS 14/23 |
£21.20
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
10189600
|
Lenz Laborglasinstrumente™
05337004 |
NS 29/32 |
£29.30
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
10366261
|
Lenz Laborglasinstrumente™
05337002 |
NS 19/26 |
£25.25
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
Description
- Tube bent at a 75° angle
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Drying Tube | |
Bent |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone