Learn More
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone
Excellent for protecting reactions against moisture while equalizing pressure. Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05337002
 View more versions of this product
Product Code. 10366261
  • £25.25 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tube bent at a 75° angle
  • Available in a range of cone sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Bent
Drying Tube
NS 19/26
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Bent Drying Tubes with NS Cone > Neck Size: 19/26