Learn More
Thermo Scientific™ Pierce™ Silver Stain Kit
Stain proteins in 1D and 2D gels with this rapid, ultra-sensitive and versatile silver stain system that yields consistent and reliable results.
Brand:  Thermo Scientific™ 24612X3
 View more versions of this product
Product Code. 30055046
  • £422.00 / 1 set
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce Silver Stain Kit is a rapid, ultra-sensitive and versatile silver stain system for protein detection in polyacrylamide gels, yielding consistent and reliable results with many 1D and 2D gel types for both first-time and experienced users.
Thermo Scientific™ Pierce Silver Stain Kit is a rapid, ultra-sensitive and versatile silver stain system for protein detection in polyacrylamide gels, yielding consistent and reliable results with many 1D and 2D gel types for both first-time and experienced users.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
In-Gel Detection
Silver
Pierce™
Protein
Colorimetric
Stain Kit
3 x 1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Silver Stain Kit > 3 x 1 L