Learn More
Invitrogen™ DTT, 1M
DTT, 1M
Brand: Invitrogen™ P2325
66.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
DTT is an essential component for kinase assays using LanthaScreen™ Technology. Invitrogen's DTT is 1M in water. It is tested for background fluorescence, making it optimal for use in high-throughput screening applications.
Specifications
DTT is shipped on dry ice and should be stored at -20°C. | |
Liquid | |
Dithiothreitol |
Dry Ice | |
1 mL |
Safety and Handling
- DTT
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : 1,4-dimercapto-2,3-Butanediol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.