missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Duotest™ pH Paper Roll
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
£13.85
Product Code | Brand | pH Range | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Graduations | Price | Quantity & Availability | ||||
10084880
|
Macherey-Nagel™
90302 |
1 to 4.3 | 1.0, 1.3, 1.6, 1.9, 2.2, 2.5, 2.8, 3.1, 3.4, 3.7, 4.0, 4.3 |
£13.85
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10320771
|
Macherey-Nagel™
90303 |
3.5 to 6.8 | 3.5, 3.8, 4.1, 4.4, 4.7, 5.0, 5.3, 5.6, 5.9, 6.2, 6.5, 6.8 |
£13.85
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10291991
|
Macherey-Nagel™
90304 |
5 to 8 | 5,0, 5,3, 5,6, 5,9, 6,2, 6,5, 6,8, 7,1, 7,4, 7,7, 8,0 |
£13.85
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360151
|
Macherey-Nagel™
90305 |
7 to 10 | 7.0, 7.3, 7.6, 7.9, 8.2, 8.5, 8.8, 9.1, 9.4, 9.7, 10.0 |
£13.85
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10094880
|
Macherey-Nagel™
90306 |
9.5 to 14 | 9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0, 13.5, 14.0 |
£13.85
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
- Provides rapid and reliable results
- Available for a range of pH gradations
- Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
- Allows more accurate reading and good estimation of intermediate values
Specifications
Roll | |
pH Paper | |
5 m | |
10 mm |