missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran Adapters with NS Stopcock

Manufactured from DURAN™ glass tubing. Lenz Laborglasinstrumente™ Duran™ Adapters with NS Stopcock includes bent glass olive (90°) and NS stopcock with solid glass plug, bore: 2.5mm.

Brand:  Lenz Laborglasinstrumente™ 05098001

Code : A0

Additional Details : Weight : 0.04500kg

 View more versions of this product

Product Code. 10567891

  • £33.55 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Manufactured from DURAN™ glass tubing
  • Includes bent glass olive (90°) and stopcock with solid glass plug, bore: 2.5mm.
Specifications

Specifications

NS 14/23
Bent glass olive (90°), Solid glass plug, bore 2.5mm
Duran Glass
Adapter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers