Learn More
DWK Life Sciences DURAN™ Beaker, high form, with spout
Due to its shape, the beaker is a universal product for a wide range of laboratory applications.
£55.65 - £270.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Heating; Diluting solutions; Mixing several components; Measuring liquid amounts |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11762232
|
DWK Life Sciences
211161704 |
50 mL |
£55.65
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11331435
|
DWK Life Sciences
211162409 |
100 mL |
£114.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11341435
|
DWK Life Sciences
211162906 |
150 mL |
£133.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11792232
|
DWK Life Sciences
211163602 |
250 mL |
£68.90
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11351435
|
DWK Life Sciences
211164101 |
400 mL |
£74.10
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11361435
|
DWK Life Sciences
211164804 |
600 mL |
£110.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11371435
|
DWK Life Sciences
211165303 |
800 mL |
£130.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11732242
|
DWK Life Sciences
211165406 |
1000 mL |
£148.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11742242
|
DWK Life Sciences
211166308 |
2000 mL |
£270.00
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11381435
|
DWK Life Sciences
211166805 |
3000 mL |
£114.00
Pack of 2 |
In Stock
Log in to see stock. |
||||||
Description
Beakers are cylindrical vessels with a straight side wall. Thanks to this shape, beakers are universally usable laboratory vessels and usually serve as cooking glasses, collecting vessels and for preparing, mixing and heating solutions. Easy filling is possible via the large vessel opening. Beakers are available in various shapes and sizes: high and low, with or without spout, with or without scale.
- Spout for clean pouring
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Borosilicate Glass (3.3) | |
Heating; Diluting solutions; Mixing several components; Measuring liquid amounts | |
Autoclavable |