Learn More
DWK Life Sciences DURAN™ Connection Piece, with standard ground cone, 90° angle
The connector allows the connection of equipment via a hose.
£15.55 - £22.95
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
10767301
|
DWK Life Sciences
243000604 |
NS 14/23 |
£15.55
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10602842
|
DWK Life Sciences
243000904 |
NS 29/32 |
£21.35
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
10203701
|
DWK Life Sciences
243000801 |
NS 24/29 |
£22.95
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||||
Description
Connectors are transition pieces with olive and have an olive instead of the second standard ground joint. Tubes can be connected via this olive, which allow e.g. the connection of a vacuum pump.
- Tight closure due to standard ground joint
- 90° angle
- With standard ground cone
- Easy and tight connection of hoses
- Fired-on, highly durable white print
Specifications
1 Ea. |