Learn More
DWK Life Sciences DURAN™ Culture Tube, with straight rim, for Kapsenberg caps
Culture tubes are used for growing cultures from liquid media.
Brand: DWK Life Sciences 261322305
Description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today's research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. The original DURAN™ culture tubes are distinguished by their uniform wall thickness distribution, minimal thermal expansion and the resulting high resistance to temperature changes. They are therefore particularly suitable for work at very high temperatures and rapid temperature changes.
- Culture tubes with straight edges can be closed with metal caps or cotton plugs to allow air exchange
- The straight rim permits the use of Kapsenberg and metal caps
Specifications
1.0 to 1.2 mm thickness | |
Borosilicate Glass (3.3) | |
100/Pk. | |
18 mm |
30 mL | |
Growth of microorganisms; For microbiological work | |
Autoclavable |