Learn More
DWK Life Sciences DURAN™ Dimroth Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
The Dimroth condenser with its internal cooling spiral has a large cooling surface and is often used as a reflux condenser.
Brand: DWK Life Sciences 242547106
89.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Due to the internal cooling spiral, the Dimroth condenser is characterized by its large cooling surface (coolant flow in counter flow principle). The most well-known application of the Dimroth condenser is the reflux condenser. A reaction mixture is heated, the resulting vapours condense on the cooling spiral and are returned to the reaction mixture.
- The Dimroth condenser comprises a coil condenser located within a tube
- This condenser type has a larger heat exchange surface and thus a better cooling effect than the Liebig or Allihn condenser
- Two removable plastic olives for inlet and outlet of the cooling medium
- Tight closure due to standard ground joint
Specifications
Product and reflux condenser; Distillation; Condensers of length 160 mm (with standard ground joint 14/23) and 250 mm (with standard ground joint 24/29 and standard ground joint 29/32) are special sizes and not according to DIN | |
NS 24/29 | |
Non-DIN Size |
250 mm | |
Borosilicate Glass (3.3), PBT, Polypropylene, Silicone |