Learn More
DWK Life Sciences DURAN™ Erlenmeyer Flask, with standard ground joint
Erlenmeyer flasks with standard ground joint allow, among other things, the storage of a solution through the tight closure of the vessel.
Brand:  DWK Life Sciences 241933204
 View more versions of this product
Product Code. 15261889
  • £224.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Due to their large footprint, Erlenmeyer flasks are suitable, for example, for preparing solutions and storing them. The additional grinding allows a tight closure of the vessel.
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white print
  • Easy mixing due to conical form
TRUSTED_SUSTAINABILITY
Specifications
Specifications
79 mm
Borosilicate Glass (3.3)
10/Pk.
NS 29/32
200 mL
Mixing several components; Diluting solutions; Measuring liquid amounts; Heating; Volume sizes 50 mL (NS 24/29), 100 mL (NS 24/29), 200 mL (NS 29/32), 250 mL (NS 45/40), 300 mL (NS 29/32), 500 mL (NS 45/40) and 1000 mL (NS 45/40) are not according to DIN ISO
Non-DIN/ISO size
Autoclavable