Learn More
DWK Life Sciences DURAN™ Evaporating Flask, with standard ground joint, pear shape
Evaporating flasks are particularly suitable for evaporating liquids or solvents.
Brand:  DWK Life Sciences 241204605
378.75 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15261859
  • £454.00 / Pack of 10
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Evaporating flasks are a variant of round flasks and have a pear-shaped shape. The standard ground joint allows a precise connection to other ground joint components, e.g. a rotary evaporator. Due to their shape, evaporating flasks allow a large uniform liquid surface, making them particularly suitable for the evaporation of solvents.
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
  • Pear shape
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Evaporation of liquids; Heating; Suitable for heating baths and mantles; Mixing several components; Rotary evaporator flask, suitable for all standard rotary evaporators; Concentration of solutions
500 mL
Borosilicate Glass (3.3)
10/Pk.
101 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Evaporating Flask, with standard ground joint, pear shape > 500 mL