Learn More
DWK Life Sciences DURAN™ Filtering Flask, with Side-Arm Socket, Erlenmeyer shape
The filtering flask is a thick-walled vessel which is used as a collecting vessel for the filtrate during filtration.
Brand:  DWK Life Sciences 211833603
Additional Details : Weight : 2.90000kg
 View more versions of this product
Product Code. 12580997
  • £368.00 / Pack of 10
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
Description
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Buechner funnels are cylinders with an integrated large-pored sieve plate. Without additional filter paper, only coarse filtration is possible. Buechner funnels can also be used in vacuum filtration. The applied vacuum accelerates the filtration.
  • Provision of filtration flasks with a socket has not only made work in preparation and analytical laboratories easier and simpler, but has also reduced the risk of accidents
  • Vacuum resistant due to the heavy wall thickness
  • Fired-on, highly durable white print
  • Erlenmeyer shape
Specifications
Specifications
85 mm
Borosilicate Glass (3.3)
10/Pk.
250 mL
Separations by vacuum filtration; Filtration at reduced pressure; Filtrieren; Vacuum filtration; Safe working under vacuum
Yes