Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser
Duran™ Glass Coil Condensers made of DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05250404
 View more versions of this product
Product Code. 10732342
  • £148.00 / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Duran™ Glass
Specifications
Specifications
DIN 12593 and in addition to DIN
For condensing vapors
1 Ea.
Duran Glass
Cooling Surface: 650 cm2
400 mm
NS 29/32
Coil Condenser