Learn More
Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads are manufactured from durable DURAN™ tubing.
Brand:  Lenz Laborglasinstrumente™ 05250214
 View more versions of this product
Product Code. 10139650
  • £123.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12593
  • With NS socket and cone
  • Corrosion-free and resistant to thermal shock
  • Water connection: GL 14 threads (with two threaded plastic olives)
Specifications
Specifications
DIN 12593 and in addition to DIN
For condensing vapors
1 Each
Duran Glass
Cooling Surface: 240cm2
160 mm
29/32
Coil Condenser