missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide cured silicone
Cap liners with PTFE faced silicone.
£8.80 - £83.20
Specifications
For Use With (Application) | Replacement cap liner for red PBT caps |
---|---|
Material | Silicone, PTFE, VMQ |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
11381475
|
DWK Life Sciences
292482806 |
GL 45 |
£83.20
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11736642
|
DWK Life Sciences
292480805 |
GL14 |
£12.30
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11746642
|
DWK Life Sciences
292481107 |
GL 18 |
£8.80
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11755434
|
DWK Life Sciences
292481304 |
GL 25 |
£14.05
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11765434
|
DWK Life Sciences
292481904 |
GL 32 |
£28.05
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
Replacement cap liners suitable for PBT screw caps and tamper-evident screw caps. Heat resistance to 130°C (steam) and 200°C (dry heat). Excellent chemical resistance due to PTFE coating. Silicon is peroxide catalysed.
- replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
- Excellent chemical resistance due to PTFE coating
- Cap liner forms a tight seal with bottle Peroxide catalysed silicone
- Peroxide catalysed silicone
- Suitable for PBT screw caps and tamper-evident screw caps
Specifications
Replacement cap liner for red PBT caps | |
Silicone, PTFE, VMQ | |
10/Pk. | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide cured silicone