Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
The screw thread coupling is used to connect two threaded connections.
Brand: DWK Life Sciences 292260556
Code : A0
Additional Details : Weight : 0.01100kg
Description
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
- For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
Specifications
Autoclavable | |
Connection of glass threads | |
GL 14 |
PBT | |
1 Ea. |