missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key

Shut-off with PTFE plug for grease free operation.

Brand:  DWK Life Sciences 286413605

 View more versions of this product

Product Code. 15854718

  • £1135.00 / Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.

  • Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
  • The 3-part tap safety catch secures the tap plug in the tap sleeve.
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 mm
Borosilicate Glass (3.3), PTFE
14.5 NS
50/Pk.
10 mm
Autoclavable
1.5 mm
Installation in glass apparatus and devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.