Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key
Shut-off with PTFE plug for grease free operation.
£15.12 - £1135.00
Specifications
Material Borosilicate Glass (3.3), PTFE
Autoclavable Autoclavable
For Use With (Application) Installation in glass apparatus and devices
View More Specs
Products 9
Product Code Brand Closure Size Plug Bore Price Quantity & Availability  
Product Code Brand Closure Size Plug Bore Price Quantity & Availability  
15884718
View Documents
DWK Life Sciences
286414601
21.5 NS 6 mm
£219.00
Pack of 5
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15844718
View Documents
DWK Life Sciences
286412806
12.5 NS 2.5 mm
£943.00
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15804728
View Documents
DWK Life Sciences
286415006
29.2 NS 10 mm
£396.00
Pack of 5
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15854718
View Documents
DWK Life Sciences
286413605
14.5 NS 4 mm
£1135.00
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15864718
View Documents
DWK Life Sciences
286413802
18.8 NS 4 mm
£736.00
Pack of 25
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15874718
View Documents
DWK Life Sciences
286414207
18.8 NS 6 mm
£777.00
Pack of 25
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15894718
View Documents
DWK Life Sciences
286414807
24 NS 8 mm
£285.00
Pack of 5
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
15834718
View Documents
DWK Life Sciences
286412609
12.5 NS 2 mm
£943.00
Pack of 50
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
10263781
View Documents
DWK Life Sciences
286413408
14.5 NS 2.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
  • Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
  • The 3-part tap safety catch secures the tap plug in the tap sleeve.
  • Taper 1:10
  • Glass articles for further processing
Specifications
Specifications
Borosilicate Glass (3.3), PTFE
Autoclavable
Installation in glass apparatus and devices