missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key
Shut-off with PTFE plug for grease free operation.
Brand: DWK Life Sciences 286413802
This item is not returnable.
View return policy
Description
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
- The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Taper 1:10
- Glass articles for further processing
Specifications
4 mm | |
Borosilicate Glass (3.3), PTFE | |
18.8 NS | |
25/Pk. |
10 mm | |
Autoclavable | |
1.5 mm | |
Installation in glass apparatus and devices |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction